2-Chloro-4-fluorotoluene, 98%
2-Chloro-4-fluorotoluene, 98%
2-Chloro-4-fluorotoluene, 98%
2-Chloro-4-fluorotoluene, 98%
Thermo Scientific Chemicals

2-Chloro-4-fluorotoluene, 98%

CAS: 452-73-3 | C7H6ClF | 144.573 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B20278.22
also known as B20278-22
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS452-73-3
IUPAC Name2-chloro-4-fluoro-1-methylbenzene
Molecular FormulaC7H6ClF
InChI KeyCSARJIQZOSVYHA-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4975-1.5015 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only