4-(4-Morpholinyl)aniline, 98+%
4-(4-Morpholinyl)aniline, 98+%
4-(4-Morpholinyl)aniline, 98+%
4-(4-Morpholinyl)aniline, 98+%
Thermo Scientific Chemicals

4-(4-Morpholinyl)aniline, 98+%

CAS: 2524-67-6 | C10H14N2O | 178.235 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20292.06
also known as B20292-06
Price (USD)
60.65
Online Exclusive
67.70
Save 7.05 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.70
Save 7.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS2524-67-6
IUPAC Name4-(morpholin-4-yl)aniline
Molecular FormulaC10H14N2O
InChI KeyPHNDZBFLOPIMSM-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pink to brown
Melting Point (clear melt)128.5-133.5?C
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

Solubility
Soluble in chloroform, ethyl acetate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
RUO – Research Use Only

General References:

  1. JRoya Esmaili; Davood Nematollahi. Electrochemical method for the synthesis of disulfides of 2-(benzo[d]thiazol(or oxazol)-2-ylamino)-5-morpholinobenzenethiol. Journal of Organic Chemistry. 2013, 78 (10), 5018-5021.
  2. Carey L Horchler; John P McCauley; James E Hall; Dean H Snyder; W Craig Moore; Thomas J Hudzik; Marc J Chapdelaine. Synthesis of novel quinolone and quinoline-2-carboxylic acid (4-morpholin-4-yl-phenyl)amides: a late-stage diversification approach to potent 5HT1B antagonists. Bioorganic & Medicinal Chemistry. 2007, 15 (2), 939-950.