Orcein, for analysis
Orcein, for analysis
Orcein, for analysis
Thermo Scientific Chemicals

Orcein, for analysis

Red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum. | CAS: 1400-62-0 | C28H24N2O7 | 500.507 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20294.03
also known as B20294-03
Price (USD)
36.65
Online Exclusive
40.50
Save 3.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.50
Save 3.85 (10%)
Each
Chemical Identifiers
CAS1400-62-0
IUPAC Name4,12-bis[(2,4-dihydroxy-6-methylphenyl)amino]-3,13-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1,3,6,9,12-pentaene-5,11-dione
Molecular FormulaC28H24N2O7
InChI KeyVPEASJIRGSVXBF-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC(O)=C1NC1=C(C)C2=C3C(OC2=CC1=O)=CC(=O)C(NC1=C(C)C=C(O)C=C1O)=C3C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying< 7.0% (110?C/const. wt.)
Color ChangepH <7.0: Red, pH >7.0 purple
lambda-max (UV-Vis)575-583nm (0.01mol/L NaOH)
Appearance (Color)Black to Brown
FormCrystals or Crystalline powder
It is a red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only