3',5'-Difluoroacetophenone, 97%
3',5'-Difluoroacetophenone, 97%
3',5'-Difluoroacetophenone, 97%
Thermo Scientific Chemicals

3',5'-Difluoroacetophenone, 97%

CAS: 123577-99-1 | C8H6F2O | 156.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number B20301.14
also known as B20301-14
Price (USD)
572.65
Special offer
Online exclusive
Ends: 30-Jun-2026
674.00
Save 101.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
572.65
Special offer
Online exclusive
Ends: 30-Jun-2026
674.00
Save 101.35 (15%)
Each
Chemical Identifiers
CAS123577-99-1
IUPAC Name1-(3,5-difluorophenyl)ethan-1-one
Molecular FormulaC8H6F2O
InChI KeyOXJLDNSPGPBDCP-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormFused solid
Melting Point (clear melt)35.0-44.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only