1-(4-Chlorophenyl)biguanide hydrochloride, 97%
1-(4-Chlorophenyl)biguanide hydrochloride, 97%
1-(4-Chlorophenyl)biguanide hydrochloride, 97%
Thermo Scientific Chemicals

1-(4-Chlorophenyl)biguanide hydrochloride, 97%

CAS: 4022-81-5 | C8H11Cl2N5 | 248.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20311.03
also known as B20311-03
Price (USD)
28.65
Online Exclusive
31.80
Save 3.15 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
31.80
Save 3.15 (10%)
Each
Chemical Identifiers
CAS4022-81-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)236.0-245.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only