4-Nitrothioanisole, 98%
4-Nitrothioanisole, 98%
4-Nitrothioanisole, 98%
Thermo Scientific Chemicals

4-Nitrothioanisole, 98%

CAS: 701-57-5 | C7H7NO2S | 169.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20339.06
also known as B20339-06
Price (USD)
36.65
Online Exclusive
41.10
Save 4.45 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
41.10
Save 4.45 (11%)
Each
Chemical Identifiers
CAS701-57-5
IUPAC Name1-(methylsulfanyl)-4-nitrobenzene
Molecular FormulaC7H7NO2S
InChI KeyNEZGPRYOJVPJKL-UHFFFAOYSA-N
SMILESCSC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)67.0-73.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only