2-Amino-2',5-dichlorobenzophenone, 98%
2-Amino-2',5-dichlorobenzophenone, 98%
2-Amino-2',5-dichlorobenzophenone, 98%
Thermo Scientific Chemicals

2-Amino-2',5-dichlorobenzophenone, 98%

CAS: 2958-36-3 | C13H9Cl2NO | 266.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number B20360.06
also known as B20360-06
Price (USD)
28.65
Online Exclusive
32.00
Save 3.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
32.00
Save 3.35 (10%)
Each
Chemical Identifiers
CAS2958-36-3
IUPAC Name4-chloro-2-(2-chlorobenzoyl)aniline
Molecular FormulaC13H9Cl2NO
InChI KeyKWZYIAJRFJVQDO-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)86.0-92.0?C
Appearance (Color)Yellow
Assay (GC)≥97.5%
2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. David L. Coffen; Beatrice Schaer; Fred T. Bizzarro; Julia B. Cheung. 2-Benzazepines. 8. Zerovalent nickel mediated biaryl synthesis of an anxiolytic pyrimido[5,4-d][2]benzazepine. J. Org. Chem. 1984, 49 (2), 296-300.
  2. Kentaro Hirai; Toshio Fujishita; Teruyuki Ishiba; Hirohiko Sugimoto; Shigeru Matsutani; Yuji Tsukinoki; Katsumi Hirose. Amino acid amide derivatives of 2-[3-(aminomethyl)-4H-1,2,4-triazol-4-yl]benzophenones, a novel class of annulated peptidoaminobenzophenones. J. Med. Chem. 1982, 25 (12), 1466-1473.