3,3-Dimethylglutaric acid, 98+%
3,3-Dimethylglutaric acid, 98+%
3,3-Dimethylglutaric acid, 98+%
Thermo Scientific Chemicals

3,3-Dimethylglutaric acid, 98+%

CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B20388.09
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Chemical Identifiers
CAS4839-46-7
IUPAC Name3,3-dimethylpentanedioic acid
Molecular FormulaC7H12O4
InChI KeyDUHQIGLHYXLKAE-UHFFFAOYSA-N
SMILESCC(C)(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or Crystalline powder
Assay (Silylated GC)≥98.0%
Solution Test(10% w/v in water) : Clear to slightly turbid
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
View more
It acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.

Solubility
Solubility in water, very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Juan Xiong, et al. Conjugates of betulin derivatives with AZT as potent anti-HIV agents.Bioorg Med Chem.2010,18(17), 6451-6469.
  2. Brendan D Kelly and Tristan H Lambert. Cyclopropenium-activated cyclodehydration of diols.Org Lett.2011,17(4), 740-743.