N-tert-Butylacrylamide, 97%
N-tert-Butylacrylamide, 97%
N-tert-Butylacrylamide, 97%
N-tert-Butylacrylamide, 97%
Thermo Scientific Chemicals

N-tert-Butylacrylamide, 97%

CAS: 107-58-4 | C7H13NO | 127.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B20394.22
also known as B20394-22
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.10
Save 9.45 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.10
Save 9.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS107-58-4
IUPAC NameN-tert-butylprop-2-enamide
Molecular FormulaC7H13NO
InChI KeyXFHJDMUEHUHAJW-UHFFFAOYSA-N
SMILESCC(C)(C)NC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
Melting Point126-132?C
FormCrystals or powder or crystalline powder and/or chunks
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only