2,4,4-Trimethylcyclopentanone, 98%
Thermo Scientific Chemicals

2,4,4-Trimethylcyclopentanone, 98%

CAS: 4694-12-6 | C8H14O | 126.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20425.03
also known as B20425-03
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.50
Save 5.85 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.50
Save 5.85 (14%)
Each
Chemical Identifiers
CAS4694-12-6
IUPAC Name2,4,4-trimethylcyclopentan-1-one
Molecular FormulaC8H14O
InChI KeyOXTQEWUBDTVSFB-UHFFFAOYNA-N
SMILESCC1CC(C)(C)CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4300-1.4330 @ 20°C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only