4-Acetamidobenzaldehyde, 98%
4-Acetamidobenzaldehyde, 98%
4-Acetamidobenzaldehyde, 98%
4-Acetamidobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Acetamidobenzaldehyde, 98%

CAS: 122-85-0 | C9H9NO2 | 163.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20430.14
also known as B20430-14
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS122-85-0
IUPAC NameN-(4-formylphenyl)acetamide
Molecular FormulaC9H9NO2
InChI KeySKLUWKYNZNXSLX-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5% (UK-sourced material)
CommentMaterial sourced in UK and US
Appearance (Color)White to cream to yellow
Melting Point (clear melt)153.0-159.0?C (UK-sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only