2,4-Difluorophenylacetic acid, 99%
2,4-Difluorophenylacetic acid, 99%
2,4-Difluorophenylacetic acid, 99%
2,4-Difluorophenylacetic acid, 99%
Thermo Scientific Chemicals

2,4-Difluorophenylacetic acid, 99%

CAS: 81228-09-3 | C8H5F2O2 | 171.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20431.06
also known as B20431-06
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS81228-09-3
IUPAC Name2-(2,4-difluorophenyl)acetate
Molecular FormulaC8H5F2O2
InChI KeyQPKZIGHNRLZBCL-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)113.0-119.0?C
Appearance (Color)White
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only