2-(2-Naphthyl)indole, 98%
2-(2-Naphthyl)indole, 98%
2-(2-Naphthyl)indole, 98%
Thermo Scientific Chemicals

2-(2-Naphthyl)indole, 98%

CAS: 23746-81-8 | C18H13N | 243.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20434.03
also known as B20434-03
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Chemical Identifiers
CAS23746-81-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)199.5-205.5?C
Appearance (Color)Pale brown
Assay (GC)≥97.5%
FormPowder
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Masaki Shimizu, et al. SEfficient blue electroluminescence of silylene-bridged 2-(2-naphthyl)indole.J. Mater. Chem.,2012,22(10), 4337-4342.
  2. Marco Bandini, et al. Catalytic enantioselective addition of indoles to arylnitroalkenes: An effective route to enantiomerically enriched tryptamine precursors.Chirality.2005,17(9), 522-529.