3-Methyl-5-hexen-3-ol, 98%
3-Methyl-5-hexen-3-ol, 98%
3-Methyl-5-hexen-3-ol, 98%
Thermo Scientific Chemicals

3-Methyl-5-hexen-3-ol, 98%

CAS: 1569-44-4 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20447.03
also known as B20447-03
Price (USD)
18.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
18.00
Each
Chemical Identifiers
CAS1569-44-4
IUPAC Name3-methylhex-5-en-3-ol
Molecular FormulaC7H14O
InChI KeyGBARKLJMQRUBKV-UHFFFAOYNA-N
SMILESCCC(C)(O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4355-1.4395 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only