1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%
1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%
1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%
Thermo Scientific Chemicals

1-Phenyl-3-trifluoromethyl-2-pyrazolin-5-one, 98%

CAS: 321-07-3 | C10H7F3N2O | 228.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20478.14
also known as B20478-14
Price (USD)
458.65
Online Exclusive
509.00
Save 50.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
458.65
Online Exclusive
509.00
Save 50.35 (10%)
Each
Chemical Identifiers
CAS321-07-3
IUPAC Name1-phenyl-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC10H7F3N2O
InChI KeyGLGRRRKQSFURGD-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NN(C(=O)C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)190.0-197.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only