1-Bromo-5-chloropentane, 98%
1-Bromo-5-chloropentane, 98%
1-Bromo-5-chloropentane, 98%
Thermo Scientific Chemicals

1-Bromo-5-chloropentane, 98%

CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20479.18
also known as B20479-18
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS54512-75-3
IUPAC Name1-bromo-5-chloropentane
Molecular FormulaC5H10BrCl
InChI KeyPHHNNDKXQVKJEP-UHFFFAOYSA-N
SMILESClCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4825-1.4865 @ 20?C
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
RUO – Research Use Only

General References:

  1. FL Lambert.; GB Ingall. Voltammetry of organic halogen compounds. IV. The reduction of organic chlorides at the vitreous (glassy) carbon electrode. Tetrahedron Letters. 197415 ( 36), 3231-3234.
  2. WA Pritts.; DG Peters. Electrochemical Reduction of 1, 5-Dihalopentanes at Carbon Cathodes in Dimethylformamide. Journal of The Electrochemical Society. 1994141 ( 12), 3318-3324.