1-Acetyl-5-bromoindoline, 98%
1-Acetyl-5-bromoindoline, 98%
1-Acetyl-5-bromoindoline, 98%
Thermo Scientific Chemicals

1-Acetyl-5-bromoindoline, 98%

CAS: 22190-38-1 | C10H10BrNO | 240.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20509.03
also known as B20509-03
Price (USD)
43.65
Online Exclusive
48.20
Save 4.55 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.20
Save 4.55 (9%)
Each
Specifications
Beilstein165052
CAS22190-38-1
Chemical Name or Material1-Acetyl-5-bromoindoline
EINECS Number000-000-0
Formula Weight240.1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only