N-Methylanthranilic acid, 90+%
N-Methylanthranilic acid, 90+%
N-Methylanthranilic acid, 90+%
N-Methylanthranilic acid, 90+%
Thermo Scientific Chemicals

N-Methylanthranilic acid, 90+%

CAS: 119-68-6 | C8H9NO2 | 151.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B20531.22
also known as B20531-22
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS119-68-6
IUPAC Name2-(methylamino)benzoic acid
Molecular FormulaC8H9NO2
InChI KeyWVMBPWMAQDVZCM-UHFFFAOYSA-N
SMILESCNC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)>90.0%
Assay (Non-aqueous acid-base Titration)>90.0% (total base)
Formpowder
Appearance (Color)Brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only