3-Amino-5-tert-butylisoxazole, 97%
3-Amino-5-tert-butylisoxazole, 97%
3-Amino-5-tert-butylisoxazole, 97%
Thermo Scientific Chemicals

3-Amino-5-tert-butylisoxazole, 97%

CAS: 55809-36-4 | C7H12N2O | 140.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20567.06
also known as B20567-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.60
Save 9.95
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.60
Save 9.95
Each
Chemical Identifiers
CAS55809-36-4
IUPAC Name5-tert-butyl-1,2-oxazol-3-amine
Molecular FormulaC7H12N2O
InChI KeyGGXGVZJHUKEJHO-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(N)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream or pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)106.5-115.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only