1-n-Butylcyclohexanol, 98%
1-n-Butylcyclohexanol, 98%
1-n-Butylcyclohexanol, 98%
Thermo Scientific Chemicals

1-n-Butylcyclohexanol, 98%

CAS: 5445-30-7 | C10H20O | 156.269 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20579.14
also known as B20579-14
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS5445-30-7
IUPAC Name1-butylcyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyRCHLXMOXBJRGNX-UHFFFAOYSA-N
SMILESCCCCC1(O)CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4620-1.4660 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only