Ethyl isonipecotate, 98%
Ethyl isonipecotate, 98%
Ethyl isonipecotate, 98%
Thermo Scientific Chemicals

Ethyl isonipecotate, 98%

CAS: 1126-09-6 | C8H15NO2 | 157.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20601.14
also known as B20601-14
Price (USD)
42.65
Online exclusive
46.70
Save 4.05
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
42.65
Online exclusive
46.70
Save 4.05
Each
Chemical Identifiers
CAS1126-09-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4580-1.4620 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Fully miscible with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Samantha J. Barry.; Richard M. Carr.; Stephen J. Lane.; William J. Leavens.; Soraya Monté.; Ian Waterhouse. Derivatisation for liquid chromatography/electrospray mass spectrometry: synthesis of pyridinium compounds and their amine and carboxylic acid derivatives.rapid communications in mass spectroscopy. 2003 , 17 (6),603-620 .
  2. Stephen A. Kolodziej.; Susan L. Hockerman.; Gary A. DeCrescenzo.; Joseph J. McDonald.; Debbie A. Mischke.; Grace E. Munie.; Theresa R. Fletcher.; Nathan Stehle.; Craig Swearingen.; Daniel P. Becker. MMP-13 selective isonipecotamide α-sulfone hydroxamates.Bioorg. Med. Chem. Lett. 2010, 20 (12),3561-3564 .