2,2-Dimethyl-3-hexanone, 99%
2,2-Dimethyl-3-hexanone, 99%
2,2-Dimethyl-3-hexanone, 99%
Thermo Scientific Chemicals

2,2-Dimethyl-3-hexanone, 99%

CAS: 5405-79-8 | C8H16O | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20612.03
also known as B20612-03
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.20
Save 7.55 (16%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.20
Save 7.55 (16%)
Each
Chemical Identifiers
CAS5405-79-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4085-1.4115 @ 20?C
Assay (GC)≥98.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only