3,4,5-Trihydroxybenzamide, 98%
3,4,5-Trihydroxybenzamide, 98%
3,4,5-Trihydroxybenzamide, 98%
3,4,5-Trihydroxybenzamide, 98%
Thermo Scientific Chemicals

3,4,5-Trihydroxybenzamide, 98%

CAS: 618-73-5 | C7H7NO4 | 169.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20620.14
also known as B20620-14
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS618-73-5
IUPAC Name3,4,5-trihydroxybenzamide
Molecular FormulaC7H7NO4
InChI KeyRBQIPEJXQPQFJX-UHFFFAOYSA-N
SMILESNC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Pale brown to gray
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only