3,4-Dimethoxyphenylacetic acid, 98%
3,4-Dimethoxyphenylacetic acid, 98%
3,4-Dimethoxyphenylacetic acid, 98%
Thermo Scientific Chemicals

3,4-Dimethoxyphenylacetic acid, 98%

CAS: 93-40-3 | C10H12O4 | 196.202 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B20625.22
also known as B20625-22
Price (USD)
86.65
Online Exclusive
95.90
Save 9.25 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
95.90
Save 9.25 (10%)
Each
Chemical Identifiers
CAS93-40-3
IUPAC Name2-(3,4-dimethoxyphenyl)acetic acid
Molecular FormulaC10H12O4
InChI KeyWUAXWQRULBZETB-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(O)=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Melting Point (clear melt)94.0-101.0?C
Appearance (Color)Pale cream
Assay (HPLC)≥97.5%
It reacts with formaldehyde in the presence of acid to give an isochromanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It reacts with formaldehyde in the presence of acid to give an isochromanone.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Arnold J. Friedhoff.; Leo E. Hollister. Comparison of the metabolism of 3,4-dimethoxyphenylethylamine and mescaline in humans. Biochem. Pharmacol. 1966, 15 (3),269-273 .
  2. Reacts with formaldehyde in the presence of acid to give an isochromanone: Org. Synth. Coll., 6, 471 (1988):