(-)-Carveol, mixture of isomers, 97%
(-)-Carveol, mixture of isomers, 97%
(-)-Carveol, mixture of isomers, 97%
Thermo Scientific Chemicals

(-)-Carveol, mixture of isomers, 97%

CAS: 99-48-9 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B20636.18
also known as B20636-18
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS99-48-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (sum of isomers)
Appearance (Color)Clear colorless to pale yellow
Optical Rotation-130 to -117°
FormLiquid
Refractive Index1.4750-1.5100 @ 20°C
It is a terpene that inhibits mammary carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a terpene that inhibits mammary carcinogenesis.

Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Carla C.C.R de Carvalho.; M.Manuela R da Fonseca. Maintenance of cell viability in the biotransformation of (-)-carveol with whole cells of Rhodococcus erythropolis. J. Mol. Catal. B: Enzym. 2002, 19-20 389-398 .
  2. Fabíola Maria N. Nunes.; M.C.C. Velosoa.; P.A. de P. Pereira.; J.B. de Andrade. Gas-phase ozonolysis of the monoterpenoids (S)-(+)-carvone, (R)-(-)-carvone, (-)-carveol, geraniol and citral. Atmos. Environ. 2005, 39 (40),7715-7730 .