4-n-Octyloxybenzaldehyde, 97%
4-n-Octyloxybenzaldehyde, 97%
4-n-Octyloxybenzaldehyde, 97%
4-n-Octyloxybenzaldehyde, 97%
Thermo Scientific Chemicals

4-n-Octyloxybenzaldehyde, 97%

CAS: 24083-13-4 | C15H22O2 | 234.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20692.06
also known as B20692-06
Price (USD)
60.65
Online Exclusive
67.70
Save 7.05 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.70
Save 7.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS24083-13-4
IUPAC Name4-(octyloxy)benzaldehyde
Molecular FormulaC15H22O2
InChI KeyKVOWZHASDIKNFK-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Free acid (titration)≤2.0%
Refractive Index1.5190-1.5240 @ 20°C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only