2,4-Diaminodiphenylamine, 98%
2,4-Diaminodiphenylamine, 98%
2,4-Diaminodiphenylamine, 98%
Thermo Scientific Chemicals

2,4-Diaminodiphenylamine, 98%

CAS: 136-17-4 | C12H13N3 | 199.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20735.06
also known as B20735-06
Price (USD)
360.65
Special offer
Online exclusive
Ends: 30-Jun-2026
424.00
Save 63.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
360.65
Special offer
Online exclusive
Ends: 30-Jun-2026
424.00
Save 63.35 (15%)
Each
Chemical Identifiers
CAS136-17-4
IUPAC NameN1-phenylbenzene-1,2,4-triamine
Molecular FormulaC12H13N3
InChI KeyVOSLIUIVGWBSOK-UHFFFAOYSA-N
SMILESNC1=CC=C(NC2=CC=CC=C2)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)126.0-133.0?C
FormPowder
Appearance (Color)Brown or purple to black
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only