3,5-Dimethylbenzo[b]thiophene, 97%
3,5-Dimethylbenzo[b]thiophene, 97%
3,5-Dimethylbenzo[b]thiophene, 97%
Thermo Scientific Chemicals

3,5-Dimethylbenzo[b]thiophene, 97%

CAS: 1964-45-0 | C10H10S | 162.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20768.03
also known as B20768-03
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.10
Save 12.45 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.10
Save 12.45 (15%)
Each
Chemical Identifiers
CAS1964-45-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Assay (GC)≥96.0%
Refractive Index1.6090-1.6140 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only