Diethylene glycol mono-n-hexyl ether, 97%
Diethylene glycol mono-n-hexyl ether, 97%
Diethylene glycol mono-n-hexyl ether, 97%
Thermo Scientific Chemicals

Diethylene glycol mono-n-hexyl ether, 97%

CAS: 112-59-4 | C10H22O3 | 190.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number B20774.0D
also known as B20774-0D
Price (USD)
480.65
Online Exclusive
534.00
Save 53.35 (10%)
Each
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)
480.65
Online Exclusive
534.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS112-59-4
IUPAC Name2-[2-(hexyloxy)ethoxy]ethan-1-ol
Molecular FormulaC10H22O3
InChI KeyGZMAAYIALGURDQ-UHFFFAOYSA-N
SMILESCCCCCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.4345-1.4395 @ 20?C
FormLiquid
Used as a solvent for coatings and degreasing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a solvent for coatings and degreasing.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. B Ballantyne and J S Vergnes. In vitro and in vivo genetic toxicology studies with diethylene glycol monohexyl ether.J Appl Toxicol.2001,21(6), 449-60.
  2. Chew CH and Gan LM. Monohexyl ether of ethylene glycol and diethylene glycol as microemulsion cosurfactants.J. Disper. Sci. Tech.;1990,11(1), 49-68.