2,2,2-Trifluoroethylamine, 98%
2,2,2-Trifluoroethylamine, 98%
2,2,2-Trifluoroethylamine, 98%
Thermo Scientific Chemicals

2,2,2-Trifluoroethylamine, 98%

CAS: 753-90-2 | C2H4F3N | 99.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20789.03
also known as B20789-03
Price (USD)
51.65
Online Exclusive
57.50
Save 5.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS753-90-2
IUPAC Name2,2,2-trifluoroethan-1-amine
Molecular FormulaC2H4F3N
InChI KeyKIPSRYDSZQRPEA-UHFFFAOYSA-N
SMILESNCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.3005-1.3045 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only