3-Nitrobenzhydrazide, 98+%
3-Nitrobenzhydrazide, 98+%
3-Nitrobenzhydrazide, 98+%
3-Nitrobenzhydrazide, 98+%
Thermo Scientific Chemicals

3-Nitrobenzhydrazide, 98+%

CAS: 618-94-0 | C7H7N3O3 | 181.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20800.06
also known as B20800-06
Price (USD)
41.65
Online Exclusive
46.30
Save 4.65 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
46.30
Save 4.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS618-94-0
IUPAC Name3-nitrobenzohydrazide
Molecular FormulaC7H7N3O3
InChI KeyNQEWXLVDAVTOHM-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)Cream
Melting Point (clear melt)149.5-155.5?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only