1,1-Cyclohexanediacetic acid, 98%
1,1-Cyclohexanediacetic acid, 98%
1,1-Cyclohexanediacetic acid, 98%
Thermo Scientific Chemicals

1,1-Cyclohexanediacetic acid, 98%

CAS: 4355-11-7 | C10H16O4 | 200.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20816.09
also known as B20816-09
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.70
Save 10.05 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.70
Save 10.05 (15%)
Each
Chemical Identifiers
CAS4355-11-7
IUPAC Name2-[1-(carboxymethyl)cyclohexyl]acetic acid
Molecular FormulaC10H16O4
InChI KeyYQPCHPBGAALCRT-UHFFFAOYSA-N
SMILESOC(=O)CC1(CC(O)=O)CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)177.0-183.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only