Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals
Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals
Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals
Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B20819.18
also known as B20819-18
50 g
Catalog number B20819.18
also known as B20819-18
Price (USD)
199.00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
199.00
Each
Add to cart
Chemical Identifiers
CAS5137-45-1
IUPAC Name1-ethoxy-2-methoxyethane
Molecular FormulaC5H12O2
InChI KeyCAQYAZNFWDDMIT-UHFFFAOYSA-N
SMILESCCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.3850-1.3890 @ 20°C
Stabilizer0.01% BHT
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. G. T. Rogers, et al. Nucleosides. Part VII. The reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.J. Chem. Soc. C,1969,(2), 203-208.
  2. A I Shatenshtein and Eduard S Petrov. Effect of the solvent on the formation and ionic dissociation of the adducts of alkali metals with aromatic hydrocarbons.Russian Chemical Reviews.,1967,36(2), 100.