2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B20882.36
also known as B20882-36
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B20882.36
also known as B20882-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS4584-49-0
IUPAC Namehydrogen (2-chloropropyl)dimethylamine chloride
Molecular FormulaC5H13Cl2N
InChI KeyOCWGRWAYARCRTQ-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(Cl)CN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale pink
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
FormPowder or lumps
Melting Point (clear melt)187.0-193.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only