3-Heptyn-1-ol, 98%
3-Heptyn-1-ol, 98%
3-Heptyn-1-ol, 98%
Thermo Scientific Chemicals

3-Heptyn-1-ol, 98%

CAS: 14916-79-1 | C7H12O | 112.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20884.06
also known as B20884-06
Price (USD)
40.65
Online Exclusive
44.90
Save 4.25 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.90
Save 4.25 (9%)
Each
Chemical Identifiers
CAS14916-79-1
IUPAC Namehept-3-yn-1-ol
Molecular FormulaC7H12O
InChI KeyPSWHODJVUOXHKA-UHFFFAOYSA-N
SMILESCCCC#CCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
Refractive Index1.4540-1.4580 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only