1H,1H,2H-Perfluoro-1-octene, 99%
1H,1H,2H-Perfluoro-1-octene, 99%
1H,1H,2H-Perfluoro-1-octene, 99%
Thermo Scientific Chemicals

1H,1H,2H-Perfluoro-1-octene, 99%

CAS: 25291-17-2 | C8H3F13 | 346.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20920.06
also known as B20920-06
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS25291-17-2
IUPAC Name3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
Molecular FormulaC8H3F13
InChI KeyFYQFWFHDPNXORA-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>98.5%
Refractive Index<1.300 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only