3,4-Dimethylbenzophenone, 99%
3,4-Dimethylbenzophenone, 99%
3,4-Dimethylbenzophenone, 99%
Thermo Scientific Chemicals

3,4-Dimethylbenzophenone, 99%

CAS: 2571-39-3 | C15H14O | 210.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20965.22
also known as B20965-22
Price (USD)
118.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
118.00
Each
Chemical Identifiers
CAS2571-39-3
IUPAC Name(3,4-dimethylphenyl)(phenyl)methanone
Molecular FormulaC15H14O
InChI KeyJENOLWCGNVWTJN-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)42.0-48.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only