N-Methylmaleic hydrazide, 97%
Thermo Scientific Chemicals

N-Methylmaleic hydrazide, 97%

CAS: 5436-01-1 | C5H6N2O2 | 126.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20975.06
also known as B20975-06
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.90
Save 13.25 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.90
Save 13.25 (14%)
Each
Chemical Identifiers
CAS5436-01-1
IUPAC Name1-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
Molecular FormulaC5H6N2O2
InChI KeyUAECOHJYXUJDOF-UHFFFAOYSA-N
SMILESCN1NC(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)213-219?C
Appearance (Color)White
FormPowder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only