1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper
1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper
1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper
1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper
Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper

CAS: 2043-57-4 | C8H4F13I | 474.004 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20984.14
also known as B20984-14
Price (USD)
116.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
116.00
Each
Add to cart
Chemical Identifiers
CAS2043-57-4
IUPAC Name1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
Molecular FormulaC8H4F13I
InChI KeyNVVZEKTVIXIUKW-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to pale yellow or pink
Assay (GC)≥96.0%
Refractive Index1.3575 - 1.3625 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
They are important intermediates in the synthesis of organic fluoride products. It has also been shown to have potential estrogenic effects.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from alkali metals, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Tsuyoshi Asakawa, et al. Micellar Pseudophase Separation Regions of 1H,1H,2H,2H-Perfluoroalkylpyridinium Chloride and Hydrocarbon Surfactants by Group Contribution Method.Langmuir.,199511(2), 478-482.
  2. Pravat Bhattacharyya, et al. Phosphorus(III) ligands with fluorous ponytails.J. Chem. Soc., Perkin Trans.,1997,24(1), 3609-3612.