4-Isobutyl-alpha-methylphenylacetic acid, 99%
4-Isobutyl-alpha-methylphenylacetic acid, 99%
4-Isobutyl-alpha-methylphenylacetic acid, 99%
4-Isobutyl-alpha-methylphenylacetic acid, 99%
Thermo Scientific Chemicals

4-Isobutyl-alpha-methylphenylacetic acid, 99%

CAS: 15687-27-1 | C13H18O2 | 206.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20989.14
also known as B20989-14
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS15687-27-1
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)75-79?C
Appearance (Color)White
Assay (HPLC)≥98.5%
FormCrystals or powder or crystalline powder
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Meade EA, et al. Differential inhibition of prostaglandin endoperoxide synthase (cyclooxygenase) isozymes by aspirin and other non-steroidal anti-inflammatory drugs.J Biol Chem.,1993268(9), 6610-4.
  2. Myers RF and Siegel MI. Differential effects of anti-inflammatory drugs on lipoxygenase and cyclo-oxygenase activities of neutrophils from a reverse passive Arthus reaction.Biochem Biophys Res Commun.1983,112(2), 586-94.