O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals
O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals
O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B21001.03
also known as B21001-03
1 g
Catalog number B21001.03
also known as B21001-03
Price (USD)
88.65
Online Exclusive
98.30
Save 9.65 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.30
Save 9.65 (10%)
Each
Chemical Identifiers
CAS41879-39-4
IUPAC NameO-(tert-butyldimethylsilyl)hydroxylamine
Molecular FormulaC6H17NOSi
InChI KeySSUCKKNRCOFUPT-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)ON
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-68.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥90.0%
Appearance (Color)Colorless
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Golebiowski A and Klopfenstein S. Solid supported synthesis of hydroxamic acids.Tetrahedron Lett.199839(21), 3397-3400.
  2. Masami Aizawa, et al. Silylation of multi-walled carbon nanotubes.Chem. Phys. Lett.2003368(1-2), 121-124.