n-Propyltrimethoxysilane, 98+%
n-Propyltrimethoxysilane, 98+%
n-Propyltrimethoxysilane, 98+%
Thermo Scientific Chemicals

n-Propyltrimethoxysilane, 98+%

CAS: 1067-25-0 | C6H16O3Si | 164.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B21033.22
also known as B21033-22
Price (USD)
119.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
119.00
Each
Chemical Identifiers
CAS1067-25-0
IUPAC Nametrimethoxy(propyl)silane
Molecular FormulaC6H16O3Si
InChI KeyHQYALQRYBUJWDH-UHFFFAOYSA-N
SMILESCCC[Si](OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
Refractive Index1.3885-1.3925 @ 20?C
FormLiquid
Identification (FTIR)Conforms
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. B. Riegel, et al. Raman spectroscopic analysis of the sol-gel processing of Full-size image (<1 K) mixtures.J. Mol. Spectrosc.1997410-411(16), 157-160.
  2. F. Bauer. Surface modification of nanoparticles for radiation curable acrylate clear coatings.Nucl. Instrum. Methods Phys. Res., Sect. B.2003208267-270.