Vinyltriethoxysilane, 97%
Vinyltriethoxysilane, 97%
Vinyltriethoxysilane, 97%
Thermo Scientific Chemicals

Vinyltriethoxysilane, 97%

CAS: 78-08-0 | C8H18O3Si | 190.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B21037.30
also known as B21037-30
Price (USD)
78.40
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
78.40
Each
Chemical Identifiers
CAS78-08-0
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.3970-1.4020 @ 20?C
FormLiquid
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with alkali, acid, water and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. O-Protected 2-hydroxyacetophenones can be ortho-alkylated in the presence of a ruthenium catalyst to give the 6-(triethoxysilylethyl) derivatives in moderate to high yields: J. Organomet. Chem., 506, 339 (1996).
  2. Trong, V. M.; Tuan, V. M.; Chinh, N. T.; Giang, N. V.; Trang, N. T. T.; Thanh, D. T. M.; Hang, T. T. X.; Hoang, T. Characterization of Fly Ash Modified with Vinyltriethoxysilane. J. Nanosci. Nanotechnol. 2015, 15 (8), 5905-5909.
  3. Oikawa, Y.; Saito, T.; Idomukai, S.; Tanaka, T.; Nishida, M.; Sawada, H. Preparation of magnesium carbonate nanoparticles encapsulated by nanocomposite material derived from fluoroalkyl end-capped vinyltrimethoxysilane oligomer-Application to the surface modification of glass and poly(methyl methacrylate). J. Fluorine. Chem. 2015, 177, 70-79.