Isobutyltrimethoxysilane, 97%
Isobutyltrimethoxysilane, 97%
Isobutyltrimethoxysilane, 97%
Thermo Scientific Chemicals

Isobutyltrimethoxysilane, 97%

CAS: 18395-30-7 | C7H18O3Si | 178.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B21048.14
also known as B21048-14
Price (USD)
25.10
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
25.10
Each
Chemical Identifiers
CAS18395-30-7
IUPAC Nametrimethoxy(2-methylpropyl)silane
Molecular FormulaC7H18O3Si
InChI KeyXYJRNCYWTVGEEG-UHFFFAOYSA-N
SMILESCO[Si](CC(C)C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3935-1.3985 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only