Chlorobenzene, 99%
Chlorobenzene, 99%
Chlorobenzene, 99%
Thermo Scientific Chemicals

Chlorobenzene, 99%

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number B21052.0F
also known as B21052-0F
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.5230-1.5260 @ 20?C
Assay (GC)≥98.5%
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Liu, M.; Han, M. and Yu, W. W. Hydrogenation of Chlorobenzene to Cyclohexane over Colloidal Pt Nanocatalysts under Ambient Conditions. Environ. Sci. Technol. 2009, 43 (7), 2519-2524.
  2. Zhang, L.; Anderson, A. W. Effect of Ozone and Sulfur Dioxide on the Photolytic Degradation of Chlorobenzene in Air. Ind. Eng. Chem. Res. 2013, 52 (9), 3315-3319.