2,4-Dinitroaniline, 99% (dry wt.), cont. up to 15% water
2,4-Dinitroaniline, 99% (dry wt.), cont. up to 15% water
2,4-Dinitroaniline, 99% (dry wt.), cont. up to 15% water
Thermo Scientific Chemicals

2,4-Dinitroaniline, 99% (dry wt.), cont. up to 15% water

CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B21080.22
also known as B21080-22
Price (USD)
44.20
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.20
Each
Chemical Identifiers
CAS97-02-9
IUPAC Name2,4-dinitroaniline
Molecular FormulaC6H5N3O4
InChI KeyLXQOQPGNCGEELI-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to pale brown
FormCrystals or powder or crystalline powder or fused/lumpy solid
Water Content (Karl Fischer Titration)≤15.0%
Melting Point (clear melt)176.0-182.0?C
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only