N-Fmoc-L-proline, 98%
N-Fmoc-L-proline, 98%
N-Fmoc-L-proline, 98%
N-Fmoc-L-proline, 98%
Thermo Scientific Chemicals

N-Fmoc-L-proline, 98%

CAS: 71989-31-6 | C20H19NO4 | 337.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21081.14
also known as B21081-14
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS71989-31-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Optical Rotation-40° ± 2° (c=1, Ethyl acetate)
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

Solubility
Soluble in methanol. (almost transparency.)

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Peter Sieber. A new acid-labile anchor group for the solid-phase synthesis of C-terminal peptide amides by the Fmoc method. Tetrahedron Lett. 1987, 28 (19),2107-2110 .
  2. Bin Zou.; Jingjun Wei.; Guorong Cai.; Dawei Ma. Synthesis of an Oxazoline Analogue of Apratoxin A. Org. Lett. 2003, 5 (19),3503-3506 .