4-Amino-4'-chlorodiphenyl ether, 97%
4-Amino-4'-chlorodiphenyl ether, 97%
4-Amino-4'-chlorodiphenyl ether, 97%
Thermo Scientific Chemicals

4-Amino-4'-chlorodiphenyl ether, 97%

CAS: 101-79-1 | C12H10ClNO | 219.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21083.09
also known as B21083-09
Price (USD)
27.65
Online Exclusive
30.10
Save 2.45 (8%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.10
Save 2.45 (8%)
Each
Chemical Identifiers
CAS101-79-1
IUPAC Name4-(4-chlorophenoxy)aniline
Molecular FormulaC12H10ClNO
InChI KeyYTISFYMPVILQRL-UHFFFAOYSA-N
SMILESNC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point97.5-106.5?C
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Y C Chui, et al. Studies on the pharmacokinetics and metabolism of 4-chlorodiphenyl ether in rats.Drug Metab Dispos.198715(1), 44-50.
  2. Livia A. Enache, et al. Development of a Scalable Synthetic Process for DG-051B, A First-in-Class Inhibitior of LTA4H.Org. Process Res. Dev.,200913(6), 1177-1184.