1,2-Bis(dichlorophosphino)ethane, 96%
1,2-Bis(dichlorophosphino)ethane, 96%
1,2-Bis(dichlorophosphino)ethane, 96%
Thermo Scientific Chemicals

1,2-Bis(dichlorophosphino)ethane, 96%

CAS: 28240-69-9 | C2H4Cl4P2 | 231.802 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21092.03
also known as B21092-03
Price (USD)
79.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
79.60
Each
Chemical Identifiers
CAS28240-69-9
IUPAC Namedichloro[2-(dichlorophosphanyl)ethyl]phosphane
Molecular FormulaC2H4Cl4P2
InChI KeySBWAJHLQMFBNIN-UHFFFAOYSA-N
SMILESClP(Cl)CCP(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay by NMR≥95.0%
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only