Bis(diphenylphosphino)methane, 97%
Bis(diphenylphosphino)methane, 97%
Bis(diphenylphosphino)methane, 97%
Thermo Scientific Chemicals

Bis(diphenylphosphino)methane, 97%

CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number B21112.04
also known as B21112-04
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Chemical Identifiers
CAS2071-20-7
IUPAC Name[(diphenylphosphanyl)methyl]diphenylphosphane
Molecular FormulaC25H22P2
InChI KeyXGCDBGRZEKYHNV-UHFFFAOYSA-N
SMILESC(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder of flakes
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)117-123?C
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. R. J. Puddephatt. Chemistry of bis(diphenylphosphino)methane.Chem. Soc. Rev.,198312(2), 99-127.
  2. Gus J. Palenik, et al. Steric vs. electronic effects in palladium-thiocyanate complexes. Crystal structures of dithiocyanato[bis(diphenylphosphino)methane](II), isothiocyanatothiocyanato[1,2-bis(diphenylphosphino)ethane]palladium(II), and diisothiocyanato[1,3-bis(diphenylphosphino)propane]palladium(II).J. Am. Chem. Soc.,197597(5), 1059-1066.